Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625400.2 | XP_005625457.1 | 3 - 4 | 506 | 167 | |
XM_022423876.1 | XP_022279584.1 | 3 - 4 | 937 | 167 | ||
XM_847138.4 | XP_852231.1 | 3 - 4 | 502 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |