Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632459.3 | XP_005632516.1 | 5 | 1607 | 116 | |
XM_005632460.3 | XP_005632517.1 | 6 | 1705 | 116 | ||
XM_014121936.2 | XP_013977411.1 | 5 | 1647 | 116 | ||
XM_022406646.1 | XP_022262354.1 | 6 | 1687 | 116 | ||
XM_022406647.1 | XP_022262355.1 | 5 | 1610 | 116 | ||
XM_022406648.1 | XP_022262356.1 | 6 | 1993 | 116 | ||
XM_022406649.1 | XP_022262357.1 | 5 | 1697 | 116 | ||
XM_541854.6 | XP_541854.1 | 5 | 1614 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |