Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623618.3 | XP_005623675.1 | 16 | 1855 | 153 | |
XM_005623619.3 | XP_005623676.1 | 16 | 1862 | 153 | ||
XM_005623620.3 | XP_005623677.1 | 15 | 1661 | 153 | ||
XM_005623621.3 | XP_005623678.1 | 14 | 1546 | 153 | ||
XM_005623622.3 | XP_005623679.1 | 16 | 1852 | 153 | ||
XM_022422189.1 | XP_022277897.1 | 9 | 1001 | 153 | ||
XM_022422190.1 | XP_022277898.1 | 9 | 886 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |