Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626849.3 | XP_005626906.1 | 8 - 9 | 1436 | 141 | |
XM_538744.6 | XP_538744.2 | 8 - 9 | 1590 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |