Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003275.3 | NP_001003275.1 | 4 | 363 | 155 | |
XM_005615729.3 | XP_005615786.1 | 3 | 313 | 155 | ||
XM_022416899.1 | XP_022272607.1 | 3 | 958 | 155 | ||
XM_022416922.1 | XP_022272630.1 | 3 | 959 | 155 | ||
GenBank mRNA | X56389.1 | - | 358 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Amplicon spans exons and probe does not span exons |