Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426585.1 | XP_022282293.1 | 676 | 198 | ||
XM_532285.5 | XP_532285.2 | 581 | 198 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |