Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022404778.1 | XP_022260486.1 | 2 - 3 | 313 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |