Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048101.2 | NP_001041566.1 | 4 | 793 | 113 | |
XM_022425733.1 | XP_022281441.1 | 3 | 524 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |