Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544475.6 | XP_544475.2 | 1 - 2 | 205 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |