Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHCGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424693.1 | XP_022280401.1 | 11 | 1769 | 151 | |
XM_022424694.1 | XP_022280402.1 | 10 | 1694 | 151 | ||
XM_022424695.1 | XP_022280403.1 | 10 | 1583 | 151 | ||
XM_022424696.1 | XP_022280404.1 | 9 | 1508 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Amplicon spans exons and probe does not span exons |