Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003290.2 | NP_001003290.1 | 3 - 4 | 398 | 87 | |
XM_022404322.1 | XP_022260030.1 | 3 - 4 | 456 | 87 | ||
GenBank mRNA | CF409774.1 | - | 3 - 4 | 473 | 87 | |
CF409775.1 | - | 1 - 2 | 453 | 87 | ||
U51644.1 | - | 1 - 2 | 161 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |