Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198658.1 | NP_001185587.1 | 252 | 128 | ||
GenBank mRNA | HM856592.1 | - | 252 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Canis familiaris |