Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_545695.3 | XP_545695.2 | 2 | 954 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |