Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624392.2 | XP_005624449.1 | 446 | 160 | ||
XM_005624394.1 | XP_005624451.1 | 365 | 160 | |||
XM_022422896.1 | XP_022278604.1 | 381 | 160 | |||
XM_022422897.1 | XP_022278605.1 | 426 | 160 | |||
XM_022422898.1 | XP_022278606.1 | 410 | 160 | |||
XM_537619.6 | XP_537619.2 | 446 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |