Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433299.4 | XP_003433347.1 | 1291 | 161 | ||
XM_005635541.3 | XP_005635598.1 | 1329 | 161 | |||
XM_534528.6 | XP_534528.2 | 1328 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |