Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621238.3 | XP_005621295.1 | 156 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |