Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616122.3 | XP_005616179.1 | 3 | 647 | 148 | |
XM_022427529.1 | XP_022283237.1 | 3 | 453 | 148 | ||
XM_849198.4 | XP_854291.1 | 3 | 533 | 148 |
Gene Symbol: LOC100855913 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638804.3 | XP_003638852.1 | 3 | 533 | 148 | |
XM_005616198.3 | XP_005616255.1 | 3 | 647 | 148 | ||
XM_022422994.1 | XP_022278702.1 | 3 | 453 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |