Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012619.1 | NP_001012637.1 | 3 - 4 | 733 | 163 | |
GenBank mRNA | AJ884915.1 | - | 3 - 4 | 733 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Canis familiaris Probe spans exons |