Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011887.2 | NP_001011887.2 | 2 | 929 | 143 | |
GenBank mRNA | AB185149.1 | - | 1 | 872 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |