Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022414584.1 | XP_022270292.1 | 7 | 1312 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |