Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013424.1 | NP_001013442.1 | 4 - 5 | 627 | 156 | |
GenBank mRNA | J02904.1 | - | 4 - 5 | 605 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |