Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284480.1 | NP_001271409.1 | 1 | 292 | 145 | |
XM_005617811.3 | XP_005617868.1 | 2 | 488 | 145 | ||
XM_022405804.1 | XP_022261512.1 | 2 | 908 | 145 | ||
XM_022405807.1 | XP_022261515.1 | 3 | 1049 | 145 | ||
GenBank mRNA | HQ709116.1 | - | 1 | 292 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |