Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_545677.6 | XP_545677.2 | 3 | 1197 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |