Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003067.2 | NP_001003067.2 | 1 | 1930 | 84 | |
GenBank mRNA | AB114672.1 | - | 1 | 1829 | 84 | |
AB114673.1 | - | 1 | 1816 | 84 | ||
AB114674.1 | - | 1 | 1823 | 84 | ||
AB114675.1 | - | 1 | 1822 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |