Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615432.3 | XP_005615489.1 | 2058 | 179 | ||
XM_005615433.3 | XP_005615490.1 | 2066 | 179 | |||
XM_005615434.3 | XP_005615491.1 | 1681 | 179 | |||
XM_022417317.1 | XP_022273025.1 | 2070 | 179 | |||
XM_022417320.1 | XP_022273028.1 | 2062 | 179 | |||
XM_022417323.1 | XP_022273031.1 | 1695 | 179 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |