Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003274.2 | NP_001003274.1 | 1 - 2 | 222 | 110 | |
GenBank mRNA | X56390.1 | - | 222 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |