Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_540085.4 | XP_540085.2 | 695 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Canis familiaris |