Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641114.3 | XP_005641171.1 | 6 - 7 | 953 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |