Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECTIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287128.1 | NP_001274057.1 | 1045 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |