Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625908.3 | XP_005625965.1 | 1 | 552 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |