Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535182.6 | XP_535182.2 | 261 | 181 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |