Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629226.3 | XP_005629283.1 | 4 | 2151 | 102 | |
XM_014119582.2 | XP_013975057.1 | 3 | 792 | 102 | ||
XM_022403316.1 | XP_022259024.1 | 3 | 796 | 102 | ||
XM_022403317.1 | XP_022259025.1 | 4 | 2069 | 102 | ||
XM_022403318.1 | XP_022259026.1 | 5 | 1648 | 102 | ||
XM_022403319.1 | XP_022259027.1 | 5 | 1260 | 102 | ||
XM_539721.6 | XP_539721.4 | 4 | 2060 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |