Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083367.1 | NP_001076836.1 | 4 | 333 | 158 | |
XM_005631051.3 | XP_005631108.1 | 5 | 513 | 158 | ||
XM_005631052.3 | XP_005631109.1 | 5 | 513 | 158 | ||
XM_022405071.1 | XP_022260779.1 | 5 | 391 | 158 | ||
GenBank mRNA | EF427641.1 | - | 4 | 333 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |