Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130437.2 | NP_001123909.2 | 5 - 6 | 741 | 84 | |
XM_005623168.3 | XP_005623225.1 | 5 - 6 | 717 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |