Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252379.1 | NP_001239308.1 | 3 - 4 | 290 | 143 | |
GenBank mRNA | DN879176.1 | - | 3 - 4 | 290 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |