Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620305.1 | XP_005620362.1 | 5190 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |