Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204842.1 | NP_001191771.1 | 2 | 2118 | 97 | |
XM_005625666.2 | XP_005625723.1 | 4 | 2356 | 97 | ||
XM_005625667.3 | XP_005625724.1 | 3 | 2234 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |