Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_537746.6 | XP_537746.2 | 2 - 3 | 413 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |