Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114679.1 | XP_013970154.1 | 1 - 2 | 116 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons |