Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628081.2 | XP_005628138.1 | 4 - 5 | 780 | 185 | |
XM_539215.5 | XP_539215.2 | 4 - 5 | 778 | 185 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |