Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618933.3 | XP_005618990.1 | 4 - 5 | 530 | 166 | |
XM_546130.6 | XP_546130.2 | 4 - 5 | 988 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |