Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107255.2 | XP_013962730.1 | 8 | 1803 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |