Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003029.1 | NP_001003029.1 | 1 | 355 | 179 | |
GenBank mRNA | AF525493.1 | - | 1 | 355 | 179 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |