Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636754.3 | XP_005636811.1 | 3 | 5172 | 91 | |
XM_022411241.1 | XP_022266949.1 | 3 | 1515 | 91 | ||
XM_022411242.1 | XP_022266950.1 | 3 | 793 | 91 | ||
XM_022411243.1 | XP_022266951.1 | 3 | 826 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |