Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631964.3 | XP_005632021.2 | 5 - 6 | 781 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |