Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424520.1 | XP_022280228.1 | 4 - 5 | 678 | 99 | |
XM_538417.5 | XP_538417.2 | 4 - 5 | 865 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons |