Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626826.3 | XP_005626883.1 | 4 | 2329 | 78 | |
XM_005626827.3 | XP_005626884.1 | 4 | 2387 | 78 | ||
XM_005626828.3 | XP_005626885.1 | 4 | 2242 | 78 | ||
XM_005626830.3 | XP_005626887.1 | 2 | 2144 | 78 | ||
XM_014117865.2 | XP_013973340.1 | 3 | 2128 | 78 | ||
XM_538734.6 | XP_538734.5 | 2 | 2184 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |