Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629309.3 | XP_005629366.1 | 4 - 5 | 502 | 112 | |
XM_014119540.2 | XP_013975015.1 | 4 - 5 | 594 | 112 | ||
XM_022403450.1 | XP_022259158.1 | 4 - 5 | 374 | 112 | ||
XM_532678.6 | XP_532678.1 | 4 - 5 | 545 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |