Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847624.5 | XP_852717.1 | 3 - 4 | 413 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |