Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618196.3 | XP_005618253.1 | 613 | 158 | ||
XM_005618197.3 | XP_005618254.1 | 599 | 158 | |||
XM_022416798.1 | XP_022272506.1 | 664 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |